Assay Design



Gene Transcripts

Gene Symbol
LRRC14
Entrez Gene ID
9684
Gene Name
leucine rich repeat containing 14
Gene Aliases
LRRC14A
Chromosome Location
Chr.8: 144517965 - 144525178 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014665.3 NP_055480.1 1 - 2 76 70
GenBank mRNA AK289473.1 - 1 - 2 44 70
D25216.1 - 1 - 2 34 70

View Full Product Details