Assay Design



Gene Transcripts

Gene Symbol
PPP6R2
Entrez Gene ID
9701
Gene Name
protein phosphatase 6 regulatory subunit 2
Gene Aliases
KIAA0685, PP6R2, SAP190, SAPS2
Chromosome Location
Chr.22: 50343317 - 50445090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242898.1 NP_001229827.1 4 - 5 795 81
NM_001242899.1 NP_001229828.1 4 - 5 795 81
NM_001242900.1 NP_001229829.1 4 - 5 795 81
NM_014678.4 NP_055493.2 4 - 5 795 81
XM_006724431.2 XP_006724494.1 5 - 6 906 81
XM_006724434.1 XP_006724497.1 6 - 7 1055 81
XM_006724435.2 XP_006724498.1 4 - 5 780 81
XM_011530720.2 XP_011529022.1 3 - 4 480 81
XM_011530721.2 XP_011529023.1 3 - 4 480 81
XM_011530722.2 XP_011529024.1 3 - 4 480 81
XM_011530723.2 XP_011529025.1 3 - 4 480 81
XM_011530724.2 XP_011529026.1 3 - 4 480 81
XM_011530726.2 XP_011529028.1 3 - 4 480 81
XM_011530727.2 XP_011529029.1 3 - 4 480 81
XM_011530728.2 XP_011529030.1 3 - 4 480 81
XM_011530729.2 XP_011529031.1 3 - 4 480 81
XM_011530730.2 XP_011529032.1 3 - 4 480 81
XM_011530731.1 XP_011529033.1 3 - 4 480 81
XM_011530732.2 XP_011529034.1 6 - 7 956 81
XM_011530734.2 XP_011529036.1 5 - 6 830 81
XM_011530735.2 XP_011529037.1 4 - 5 780 81
XM_011530736.2 XP_011529038.1 5 - 6 906 81
XM_011530737.2 XP_011529039.1 6 - 7 1055 81
XM_011530738.2 XP_011529040.1 5 - 6 858 81
XM_011530739.2 XP_011529041.1 3 - 4 520 81
XM_011530740.2 XP_011529042.1 5 - 6 928 81
XM_017029116.1 XP_016884605.1 3 - 4 480 81
XM_017029117.1 XP_016884606.1 7 - 8 1105 81
XM_017029118.1 XP_016884607.1 7 - 8 1057 81
XM_017029119.1 XP_016884608.1 6 - 7 979 81
XM_017029120.1 XP_016884609.1 6 - 7 956 81
XM_017029121.1 XP_016884610.1 4 - 5 780 81
XM_017029122.1 XP_016884611.1 5 - 6 858 81
XM_017029123.1 XP_016884612.1 4 - 5 780 81
XM_017029124.1 XP_016884613.1 5 - 6 906 81
XM_017029125.1 XP_016884614.1 4 - 5 780 81
XM_017029126.1 XP_016884615.1 4 - 5 780 81
XM_017029127.1 XP_016884616.1 5 - 6 858 81
XM_017029128.1 XP_016884617.1 5 - 6 906 81
XM_017029129.1 XP_016884618.1 4 - 5 780 81
XM_017029130.1 XP_016884619.1 4 - 5 780 81
XM_017029131.1 XP_016884620.1 5 - 6 906 81
XM_017029132.1 XP_016884621.1 5 - 6 858 81
XM_017029133.1 XP_016884622.1 4 - 5 734 81
XM_017029134.1 XP_016884623.1 4 - 5 734 81
XM_017029135.1 XP_016884624.1 5 - 6 784 81
XM_017029136.1 XP_016884625.1 4 - 5 734 81
XM_017029137.1 XP_016884626.1 4 - 5 734 81
XM_017029138.1 XP_016884627.1 4 - 5 734 81
GenBank mRNA AB014585.1 - 4 - 5 767 81
AK302472.1 - 4 - 5 706 81
BC000976.4 - 5 - 6 784 81
BC006568.2 - 4 - 5 781 81
BC032664.1 - 4 - 5 792 81
BC041698.1 - 4 - 5 762 81
BC052995.2 - 4 - 5 701 81

View Full Product Details