Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014713.4 | NP_055528.1 | 4 - 5 | 942 | 76 |
GenBank mRNA | AK129875.1 | - | 4 - 5 | 592 | 76 |
AY359028.1 | - | 4 - 5 | 558 | 76 | |
BC000421.2 | - | 4 - 5 | 519 | 76 | |
BC003158.1 | - | 4 - 5 | 582 | 76 | |
BC093075.1 | - | 4 - 5 | 542 | 76 | |
CR456758.1 | - | 4 - 5 | 434 | 76 | |
D14696.1 | - | 4 - 5 | 580 | 76 |