Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135214.1 | NP_001128686.1 | 7 - 8 | 881 | 115 |
NM_014776.3 | NP_055591.2 | 7 - 8 | 881 | 115 | |
NM_057169.3 | NP_476510.1 | 7 - 8 | 881 | 115 | |
NM_057170.3 | NP_476511.1 | 7 - 8 | 881 | 115 | |
NM_139201.2 | NP_631940.2 | 7 - 8 | 881 | 115 | |
XM_006719707.3 | XP_006719770.1 | 7 - 8 | 876 | 115 | |
XM_006719708.3 | XP_006719771.1 | 7 - 8 | 876 | 115 | |
XM_006719709.3 | XP_006719772.1 | 7 - 8 | 876 | 115 | |
XM_006719712.3 | XP_006719775.1 | 7 - 8 | 876 | 115 | |
XM_006719713.3 | XP_006719776.1 | 7 - 8 | 873 | 115 | |
XM_006719714.3 | XP_006719777.1 | 7 - 8 | 876 | 115 | |
XM_017020259.1 | XP_016875748.1 | 7 - 8 | 873 | 115 | |
XM_017020260.1 | XP_016875749.1 | 7 - 8 | 873 | 115 | |
GenBank mRNA | AB451294.1 | - | 7 - 8 | 716 | 115 |
AB451428.1 | - | 7 - 8 | 716 | 115 | |
AF124491.1 | - | 7 - 8 | 716 | 115 | |
AK222736.1 | - | 7 - 8 | 788 | 115 | |
AK303198.1 | - | 6 - 7 | 582 | 115 | |
AK304240.1 | - | 7 - 8 | 865 | 115 | |
AK308304.1 | - | 7 - 8 | 796 | 115 | |
BC001379.2 | - | 7 - 8 | 781 | 115 | |
BC014223.2 | - | 6 - 7 | 632 | 115 | |
BT007312.1 | - | 7 - 8 | 716 | 115 | |
CR533568.1 | - | 7 - 8 | 716 | 115 |