Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014808.3 | NP_055623.1 | 18 - 19 | 2298 | 88 |
XM_005247050.3 | XP_005247107.1 | 18 - 19 | 2262 | 88 | |
XM_011512233.1 | XP_011510535.1 | 18 - 19 | 2262 | 88 | |
XM_011512234.1 | XP_011510536.1 | 18 - 19 | 2187 | 88 | |
XM_011512235.1 | XP_011510537.1 | 19 - 20 | 2520 | 88 | |
XM_011512236.1 | XP_011510538.1 | 18 - 19 | 2316 | 88 | |
XM_017005416.1 | XP_016860905.1 | 19 - 20 | 2477 | 88 | |
XM_017005417.1 | XP_016860906.1 | 19 - 20 | 2477 | 88 | |
XM_017005418.1 | XP_016860907.1 | 16 - 17 | 2115 | 88 | |
GenBank mRNA | AB018336.1 | - | 18 - 19 | 2251 | 88 |