Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286527.2 | NP_001273456.2 | 11 - 12 | 1078 | 101 |
NM_001291454.1 | NP_001278383.1 | 8 - 9 | 658 | 101 | |
NM_014861.3 | NP_055676.3 | 11 - 12 | 1078 | 101 | |
XM_011523486.2 | XP_011521788.1 | 11 - 12 | 1168 | 101 | |
GenBank mRNA | AB014603.1 | - | 9 - 10 | 1476 | 101 |
AK091051.1 | - | 8 - 9 | 658 | 101 | |
AK300526.1 | - | 11 - 12 | 1075 | 101 | |
AK316128.1 | - | 11 - 12 | 1063 | 101 | |
AY791884.1 | - | 11 - 12 | 1074 | 101 | |
BX648333.1 | - | 11 - 12 | 1075 | 101 | |
CR749829.1 | - | 11 - 12 | 1049 | 101 |