Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014868.4 | NP_055683.3 | 14 - 15 | 2629 | 80 |
XM_005254013.2 | XP_005254070.1 | 15 - 16 | 2721 | 80 | |
XM_006719717.2 | XP_006719780.1 | 14 - 15 | 2664 | 80 | |
XM_017020282.1 | XP_016875771.1 | 15 - 16 | 2706 | 80 | |
GenBank mRNA | AB027196.1 | - | 14 - 15 | 2593 | 80 |
AK094011.1 | - | 15 - 16 | 2530 | 80 | |
AK097807.1 | - | 15 - 16 | 2555 | 80 | |
AK126274.1 | - | 15 - 16 | 2434 | 80 | |
AK301608.1 | - | 15 - 16 | 2503 | 80 | |
AK315558.1 | - | 14 - 15 | 2611 | 80 | |
AL389976.1 | - | 13 - 14 | 1901 | 80 | |
BC016622.1 | - | 14 - 15 | 2593 | 80 | |
BC031596.1 | - | 14 - 15 | 2593 | 80 | |
BC101709.1 | - | 14 - 15 | 2219 | 80 | |
BC101715.1 | - | 14 - 15 | 2219 | 80 | |
BT006974.1 | - | 14 - 15 | 2146 | 80 | |
D87451.2 | - | 14 - 15 | 2146 | 80 |