Assay Design



Gene Transcripts

Gene Symbol
NLGN1
Entrez Gene ID
22871
Gene Name
neuroligin 1
Gene Aliases
NL1
Chromosome Location
Chr.3: 173395899 - 174291287 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014932.3 NP_055747.1 5 - 6 1287 70
XM_005247234.2 XP_005247291.1 4 - 5 1462 70
XM_005247235.3 XP_005247292.1 4 - 5 1658 70
XM_005247237.3 XP_005247294.1 3 - 4 1956 70
XM_006713540.3 XP_006713603.1 5 - 6 1718 70
XM_011512551.2 XP_011510853.1 6 - 7 1569 70
XM_011512552.2 XP_011510854.1 5 - 6 2005 70
XM_011512553.2 XP_011510855.1 2 - 3 358 70
XM_017005888.1 XP_016861377.1 5 - 6 1866 70
XM_017005889.1 XP_016861378.1 5 - 6 1866 70
XM_017005890.1 XP_016861379.1 5 - 6 1522 70
XM_017005891.1 XP_016861380.1 5 - 6 1718 70
XM_017005892.1 XP_016861381.1 5 - 6 2020 70
XM_017005893.1 XP_016861382.1 6 - 7 2132 70
XM_017005894.1 XP_016861383.1 5 - 6 3650 70
XM_017005895.1 XP_016861384.1 5 - 6 7823 70
XM_017005896.1 XP_016861385.1 5 - 6 1304 70
XM_017005898.1 XP_016861387.1 4 - 5 1806 70
XM_017005899.1 XP_016861388.1 4 - 5 1945 70
XM_017005900.1 XP_016861389.1 5 - 6 1451 70
XM_017005901.1 XP_016861390.1 4 - 5 1507 70
XM_017005905.1 XP_016861394.1 2 - 3 1649 70
GenBank mRNA AB028993.1 - 4 - 5 1527 70
AK307813.1 - 6 - 7 1413 70
BC032555.1 - 5 - 6 1292 70
GQ489206.1 - 5 - 6 978 70

View Full Product Details