Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015140.3 | NP_055955.1 | 11 - 12 | 1641 | 86 |
GenBank mRNA | BC001070.2 | - | 11 - 12 | 1605 | 86 |
CT841516.1 | - | 11 - 12 | 1582 | 86 | |
D63487.1 | - | 11 - 12 | 1563 | 86 |