Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015332.3 | NP_056147.2 | 2 - 3 | 698 | 74 |
XM_011515247.2 | XP_011513549.1 | 2 - 3 | 787 | 74 | |
XM_017011908.1 | XP_016867397.1 | 2 - 3 | 787 | 74 | |
GenBank mRNA | AB028991.1 | - | 2 - 3 | 492 | 74 |
AK024226.1 | - | 1 - 2 | 206 | 74 | |
BC003691.2 | - | 2 - 3 | 559 | 74 | |
BC011673.2 | - | 1 - 2 | 318 | 74 | |
BC035014.2 | - | 2 - 3 | 596 | 74 | |
EU350932.1 | - | 2 - 3 | 516 | 74 |