Assay Design
Gene Transcripts
- Gene Symbol
- FAM65B
- Entrez Gene ID
-
9750
- Gene Name
- family with sequence similarity 65 member B
- Gene Aliases
-
C6orf32, DFNB104, DIFF40, DIFF48, MYONAP, PL48
- Chromosome Location
-
Chr.6: 24804281 - 25042288 on Build GRCh38
View Full Product Details