Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016199.2 | NP_057283.1 | 3 - 4 | 220 | 61 |
XM_017026869.1 | XP_016882358.1 | 4 - 5 | 363 | 61 | |
GenBank mRNA | AF182293.1 | - | 2 - 3 | 180 | 61 |
BC018621.1 | - | 2 - 3 | 173 | 61 | |
BI553911.1 | - | 3 - 4 | 224 | 61 |