Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016287.3 | NP_057371.2 | 9 - 10 | 1120 | 85 |
XM_005245875.4 | XP_005245932.1 | 9 - 10 | 1695 | 85 | |
XM_005245876.3 | XP_005245933.1 | 9 - 10 | 1228 | 85 | |
XM_005245877.4 | XP_005245934.1 | 9 - 10 | 1556 | 85 | |
XM_005245878.4 | XP_005245935.1 | 9 - 10 | 1492 | 85 | |
XM_005245879.4 | XP_005245936.1 | 8 - 9 | 1019 | 85 | |
XM_011541532.1 | XP_011539834.1 | 9 - 10 | 1212 | 85 | |
XM_011541533.1 | XP_011539835.1 | 9 - 10 | 1409 | 85 | |
XM_011541534.2 | XP_011539836.1 | 8 - 9 | 1029 | 85 | |
XM_017001393.1 | XP_016856882.1 | 9 - 10 | 1175 | 85 | |
XM_017001394.1 | XP_016856883.1 | 9 - 10 | 1540 | 85 | |
GenBank mRNA | AF113534.1 | - | 8 - 9 | 1053 | 85 |
AK023129.1 | - | 9 - 10 | 1120 | 85 | |
AK291252.1 | - | 9 - 10 | 1098 | 85 | |
AK304065.1 | - | 8 - 9 | 996 | 85 | |
AL833978.1 | - | 5 - 6 | 936 | 85 | |
BC045660.1 | - | 5 - 6 | 1752 | 85 |