Assay Design



Gene Transcripts

Gene Symbol
ARPP21
Entrez Gene ID
10777
Gene Name
cAMP regulated phosphoprotein 21
Gene Aliases
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location
Chr.3: 35638981 - 35798933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025068.1 NP_001020239.1 2 - 3 246 75
NM_001025069.1 NP_001020240.1 2 - 3 255 75
NM_001267616.1 NP_001254545.1 2 - 3 1402 75
NM_001267617.1 NP_001254546.1 2 - 3 212 75
NM_001267618.1 NP_001254547.1 1 - 2 343 75
NM_001267619.1 NP_001254548.1 1 - 2 177 75
NM_016300.4 NP_057384.2 2 - 3 229 75
XM_005264811.2 XP_005264868.1 2 - 3 258 75
XM_006712943.2 XP_006713006.1 2 - 3 258 75
XM_006712944.2 XP_006713007.1 2 - 3 258 75
XM_011533298.2 XP_011531600.1 2 - 3 258 75
XM_011533299.2 XP_011531601.1 2 - 3 802 75
XM_011533300.2 XP_011531602.1 2 - 3 421 75
XM_011533301.2 XP_011531603.1 2 - 3 252 75
XM_017005574.1 XP_016861063.1 2 - 3 348 75
XM_017005576.1 XP_016861065.1 2 - 3 802 75
XM_017005577.1 XP_016861066.1 2 - 3 252 75
XM_017005578.1 XP_016861067.1 2 - 3 228 75
XM_017005579.1 XP_016861068.1 2 - 3 802 75
XM_017005582.1 XP_016861071.1 2 - 3 802 75
XM_017005583.1 XP_016861072.1 2 - 3 252 75
XM_017005584.1 XP_016861073.1 2 - 3 802 75
XM_017005585.1 XP_016861074.1 2 - 3 802 75
XM_017005586.1 XP_016861075.1 2 - 3 258 75
XM_017005587.1 XP_016861076.1 2 - 3 421 75
XM_017005591.1 XP_016861080.1 2 - 3 802 75
XM_017005592.1 XP_016861081.1 2 - 3 802 75
XM_017005593.1 XP_016861082.1 2 - 3 348 75
XM_017005594.1 XP_016861083.1 2 - 3 252 75
XM_017005596.1 XP_016861085.1 2 - 3 802 75
XM_017005597.1 XP_016861086.1 2 - 3 802 75
XM_017005599.1 XP_016861088.1 2 - 3 802 75
XM_017005600.1 XP_016861089.1 2 - 3 421 75
XM_017005601.1 XP_016861090.1 2 - 3 252 75
XM_017005602.1 XP_016861091.1 2 - 3 802 75
XM_017005603.1 XP_016861092.1 2 - 3 421 75
XM_017005604.1 XP_016861093.1 2 - 3 252 75
XM_017005607.1 XP_016861096.1 2 - 3 802 75
XM_017005608.1 XP_016861097.1 2 - 3 802 75
XM_017005610.1 XP_016861099.1 2 - 3 802 75
XM_017005611.1 XP_016861100.1 2 - 3 802 75
XM_017005612.1 XP_016861101.1 2 - 3 802 75
GenBank mRNA AF112220.1 - 1 - 2 202 75
AK055064.1 - 2 - 3 255 75
AK289649.1 - 1 - 2 192 75
AK289868.1 - 2 - 3 249 75
AK294484.1 - 1 - 2 190 75
AV718064.1 - 2 - 3 246 75
AV718166.1 - 2 - 3 255 75
BC036399.1 - 2 - 3 217 75
BC041385.1 - 2 - 3 990 75
BC051828.1 - 2 - 3 229 75
BX488342.1 - 1 - 2 341 75
DA275579.1 - 1 - 2 343 75

View Full Product Details