Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017509.3 | NP_059979.2 | 1 - 2 | 78 | 69 |
XM_006723265.3 | XP_006723328.1 | 2 - 3 | 879 | 69 | |
XM_011527087.2 | XP_011525389.1 | 2 - 3 | 883 | 69 | |
XM_011527088.2 | XP_011525390.1 | 2 - 3 | 889 | 69 | |
XM_017026943.1 | XP_016882432.1 | 2 - 3 | 889 | 69 | |
GenBank mRNA | AY373373.1 | - | 1 - 2 | 47 | 69 |
AY373374.1 | - | 1 - 2 | 47 | 69 | |
BC069507.1 | - | 1 - 2 | 78 | 69 | |
BC126137.1 | - | 1 - 2 | 68 | 69 |