Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017581.3 | NP_060051.2 | 3 - 4 | 503 | 69 |
GenBank mRNA | AF227732.1 | - | 3 - 4 | 346 | 69 |
AJ243342.1 | - | 3 - 4 | 380 | 69 | |
BC113549.1 | - | 3 - 4 | 459 | 69 | |
BC113575.1 | - | 3 - 4 | 459 | 69 |