Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195248.1 | NP_001182177.1 | 6 - 7 | 846 | 57 |
NM_001195249.1 | NP_001182178.1 | 6 - 7 | 1015 | 57 | |
NM_001195250.1 | NP_001182179.1 | 5 - 6 | 684 | 57 | |
NM_001195251.1 | NP_001182180.1 | 7 - 8 | 952 | 57 | |
NM_001195252.1 | NP_001182181.1 | 6 - 7 | 630 | 57 | |
NM_001195254.1 | NP_001182183.1 | 5 - 6 | 706 | 57 | |
NM_175069.2 | NP_778239.1 | 6 - 7 | 846 | 57 | |
NM_175073.2 | NP_778243.1 | 7 - 8 | 952 | 57 | |
NR_036576.1 | 6 - 7 | 841 | 57 | ||
NR_036577.1 | 6 - 7 | 731 | 57 | ||
NR_036578.1 | 6 - 7 | 863 | 57 | ||
NR_036579.1 | 6 - 7 | 1010 | 57 | ||
XM_006716791.3 | XP_006716854.1 | 6 - 7 | 854 | 57 | |
XM_006716792.2 | XP_006716855.1 | 6 - 7 | 872 | 57 | |
XM_011517938.1 | XP_011516240.1 | 5 - 6 | 830 | 57 | |
XM_011517939.2 | XP_011516241.1 | 5 - 6 | 785 | 57 | |
XM_017014837.1 | XP_016870326.1 | 5 - 6 | 935 | 57 | |
XM_017014838.1 | XP_016870327.1 | 6 - 7 | 828 | 57 | |
GenBank mRNA | AB603741.1 | - | 5 - 6 | 742 | 57 |
AK000164.1 | - | 5 - 6 | 849 | 57 | |
AK055672.1 | - | 6 - 7 | 833 | 57 | |
AK131046.1 | - | 6 - 7 | 798 | 57 | |
AY040777.1 | - | 6 - 7 | 821 | 57 | |
AY208829.1 | - | 6 - 7 | 821 | 57 | |
AY208830.1 | - | 5 - 6 | 684 | 57 | |
AY208831.1 | - | 6 - 7 | 596 | 57 | |
AY208832.1 | - | 5 - 6 | 659 | 57 | |
AY208833.1 | - | 5 - 6 | 659 | 57 | |
AY208834.1 | - | 5 - 6 | 760 | 57 | |
AY208835.1 | - | 6 - 7 | 801 | 57 | |
AY208836.1 | - | 6 - 7 | 961 | 57 | |
AY208837.1 | - | 7 - 8 | 918 | 57 | |
AY208838.1 | - | 5 - 6 | 777 | 57 | |
AY208839.1 | - | 6 - 7 | 955 | 57 | |
AY208840.1 | - | 6 - 7 | 818 | 57 | |
AY208841.1 | - | 5 - 6 | 655 | 57 | |
AY208842.1 | - | 3 - 4 | 455 | 57 | |
AY302067.1 | - | 5 - 6 | 964 | 57 | |
AY302068.1 | - | 5 - 6 | 959 | 57 | |
AY302069.1 | - | 5 - 6 | 849 | 57 | |
AY302070.1 | - | 5 - 6 | 844 | 57 | |
BC001628.1 | - | 5 - 6 | 669 | 57 | |
BC032650.1 | - | 6 - 7 | 887 | 57 | |
BC068107.1 | - | 7 - 8 | 927 | 57 | |
BC104881.1 | - | 6 - 7 | 860 | 57 | |
BX538161.1 | - | 7 - 8 | 931 | 57 |