Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017738.3 | NP_060208.2 | 15 - 16 | 2688 | 60 |
XM_006716793.3 | XP_006716856.1 | 15 - 16 | 2486 | 60 | |
XM_011517941.2 | XP_011516243.1 | 15 - 16 | 2688 | 60 | |
XM_017014839.1 | XP_016870328.1 | 15 - 16 | 2688 | 60 | |
XM_017014840.1 | XP_016870329.1 | 15 - 16 | 2688 | 60 | |
XM_017014841.1 | XP_016870330.1 | 14 - 15 | 2493 | 60 | |
XM_017014842.1 | XP_016870331.1 | 15 - 16 | 2688 | 60 | |
XM_017014843.1 | XP_016870332.1 | 15 - 16 | 2688 | 60 | |
XM_017014844.1 | XP_016870333.1 | 8 - 9 | 1364 | 60 | |
XM_017014845.1 | XP_016870334.1 | 6 - 7 | 2169 | 60 | |
XM_017014846.1 | XP_016870335.1 | 3 - 4 | 692 | 60 | |
XM_017014847.1 | XP_016870336.1 | 3 - 4 | 685 | 60 | |
GenBank mRNA | AK000283.1 | - | 11 - 12 | 1783 | 60 |
BX647069.1 | - | 15 - 16 | 2647 | 60 |