Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018000.2 | NP_060470.2 | 2 - 3 | 551 | 68 |
GenBank mRNA | AF068290.1 | - | 1 - 2 | 50 | 68 |
AK000978.1 | - | 2 - 3 | 521 | 68 | |
BC032747.1 | - | 2 - 3 | 560 | 68 | |
BC082990.1 | - | 2 - 3 | 541 | 68 | |
BQ928130.1 | - | 2 - 3 | 551 | 68 | |
CX781801.1 | - | 2 - 3 | 546 | 68 |