Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134387.1 | NP_001127859.1 | 2 - 3 | 444 | 82 |
NM_001134388.1 | NP_001127860.1 | 3 - 4 | 353 | 82 | |
NM_001134389.1 | NP_001127861.1 | 2 - 3 | 222 | 82 | |
NM_018106.3 | NP_060576.1 | 2 - 3 | 287 | 82 | |
XM_005249793.2 | XP_005249850.1 | 3 - 4 | 397 | 82 | |
GenBank mRNA | AF201931.1 | - | 1 - 2 | 216 | 82 |
AK001341.1 | - | 2 - 3 | 203 | 82 | |
AK223532.1 | - | 2 - 3 | 287 | 82 | |
AK308971.1 | - | 2 - 3 | 202 | 82 | |
AL136674.1 | - | 2 - 3 | 195 | 82 | |
AY359090.1 | - | 2 - 3 | 375 | 82 | |
BC001239.1 | - | 3 - 4 | 306 | 82 | |
BQ925460.1 | - | 3 - 4 | 313 | 82 | |
CN365375.1 | - | 2 - 3 | 403 | 82 |