Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018141.3 | NP_060611.2 | 1 - 2 | 93 | 106 |
XM_005249206.2 | XP_005249263.1 | 1 - 2 | 105 | 106 | |
XM_017010996.1 | XP_016866485.1 | 1 - 2 | 68 | 106 | |
GenBank mRNA | AK001429.1 | - | 1 - 2 | 106 | |
BG776014.1 | - | 1 - 2 | 94 | 106 |