Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018154.2 | NP_060624.1 | 2 - 3 | 401 | 78 |
GenBank mRNA | AB104486.1 | - | 2 - 3 | 229 | 78 |
AF279307.1 | - | 2 - 3 | 322 | 78 | |
AK001288.1 | - | 2 - 3 | 360 | 78 | |
AK001466.1 | - | 2 - 3 | 392 | 78 | |
AK223080.1 | - | 2 - 3 | 355 | 78 | |
AK302134.1 | - | 2 - 3 | 362 | 78 | |
BC007726.2 | - | 2 - 3 | 325 | 78 | |
BC010014.2 | - | 2 - 3 | 335 | 78 | |
BC036521.1 | - | 2 - 3 | 365 | 78 | |
CR457235.1 | - | 2 - 3 | 229 | 78 |