Assay Design



Gene Transcripts

Gene Symbol
BTBD7
Entrez Gene ID
55727
Gene Name
BTB domain containing 7
Gene Aliases
FUP1
Chromosome Location
Chr.14: 93237550 - 93333092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018167.4 NP_060637.1 3 - 4 1537 68
GenBank mRNA AK001510.1 - 3 - 4 1463 68
BC047071.1 - 3 - 4 1534 68
BC141850.1 - 3 - 4 1478 68

View Full Product Details