Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018207.2 | NP_060677.2 | 1 - 2 | 1051 | 68 |
XM_017001629.1 | XP_016857118.1 | 2 - 3 | 901 | 68 | |
XM_017001630.1 | XP_016857119.1 | 2 - 3 | 956 | 68 | |
XM_017001631.1 | XP_016857120.1 | 2 - 3 | 3183 | 68 | |
GenBank mRNA | AK001621.1 | - | 1 - 2 | 647 | 68 |
AK122896.1 | - | 1 - 2 | 1031 | 68 | |
AX775831.1 | - | 1 - 2 | 455 | 68 | |
BC001222.1 | - | 1 - 2 | 446 | 68 | |
BC007999.1 | - | 1 - 2 | 446 | 68 | |
BC011689.2 | - | 1 - 2 | 446 | 68 | |
BC012152.1 | - | 1 - 2 | 446 | 68 |