Assay Design



Gene Transcripts

Gene Symbol
TBX4
Entrez Gene ID
9496
Gene Name
T-box 4
Gene Aliases
ICPPS, SPS
Chromosome Location
Chr.17: 61452418 - 61485110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018488.3 NP_060958.2 7 - 8 1025 90
XM_011525491.2 XP_011523793.1 8 - 9 1223 90
GenBank mRNA AF188703.1 - 7 - 8 1067 90
BC136403.1 - 8 - 9 1049 90

View Full Product Details