Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287586.1 | NP_001274515.1 | 2 - 3 | 285 | 83 |
NM_001287587.1 | NP_001274516.1 | 3 - 4 | 402 | 83 | |
NM_018845.3 | NP_061333.2 | 3 - 4 | 382 | 83 | |
XM_005245339.4 | XP_005245396.1 | 3 - 4 | 568 | 83 | |
XM_005245340.3 | XP_005245397.1 | 3 - 4 | 649 | 83 | |
XM_005245341.4 | XP_005245398.1 | 3 - 4 | 600 | 83 | |
XM_005245342.3 | XP_005245399.1 | 2 - 3 | 208 | 83 | |
XM_006711453.2 | XP_006711516.1 | 3 - 4 | 382 | 83 | |
GenBank mRNA | AF126023.1 | - | 3 - 4 | 342 | 83 |
AK289551.1 | - | 3 - 4 | 383 | 83 | |
BC005943.1 | - | 3 - 4 | 326 | 83 | |
BP248307.1 | - | 3 - 4 | 527 | 83 | |
BU160680.1 | - | 3 - 4 | 324 | 83 | |
CR457148.1 | - | 3 - 4 | 282 | 83 | |
DA113832.1 | - | 3 - 4 | 383 | 83 |