Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 34363911-34364453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNE2
Entrez Gene ID
9992
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 2
Gene Aliases
ATFB4, LQT5, LQT6, MIRP1
UniGene
Hs.551521
Gene Chromosome Location
Chr.21: 34364024-34371141 on Build GRCh38
Transcripts
NM_172201.1

SNP Detail

SNP ID Chromosome Location
rs786205806 Chr.21: 34364156-34364156 on Build GRCh38
rs971327228 Chr.21: 34363948-34363948 on Build GRCh38
rs1040354347 Chr.21: 34364042-34364042 on Build GRCh38
rs775470229 Chr.21: 34364322-34364322 on Build GRCh38
rs566035575 Chr.21: 34363938-34363938 on Build GRCh38
rs537511302 Chr.21: 34364253-34364253 on Build GRCh38
rs922487534 Chr.21: 34364046-34364046 on Build GRCh38
rs764708339 Chr.21: 34364302-34364302 on Build GRCh38
rs966106103 Chr.21: 34364369-34364369 on Build GRCh38
rs983132329 Chr.21: 34364411-34364411 on Build GRCh38
rs759027023 Chr.21: 34364145-34364145 on Build GRCh38
rs146989686 Chr.21: 34364311-34364311 on Build GRCh38
rs1033170659 Chr.21: 34364207-34364207 on Build GRCh38
rs538932486 Chr.21: 34364390-34364390 on Build GRCh38
rs931898854 Chr.21: 34364129-34364129 on Build GRCh38
rs919278114 Chr.21: 34364375-34364375 on Build GRCh38
rs957460845 Chr.21: 34363946-34363946 on Build GRCh38
rs999538522 Chr.21: 34364200-34364200 on Build GRCh38
rs765082988 Chr.21: 34364197-34364197 on Build GRCh38
rs566735365 Chr.21: 34364084-34364084 on Build GRCh38
rs944747362 Chr.21: 34363997-34363997 on Build GRCh38
rs929891264 Chr.21: 34364402-34364402 on Build GRCh38
rs910546844 Chr.21: 34363977-34363977 on Build GRCh38
rs567055431 Chr.21: 34364174-34364174 on Build GRCh38
rs986650503 Chr.21: 34363972-34363972 on Build GRCh38
rs976024402 Chr.21: 34364374-34364374 on Build GRCh38
rs569123981 Chr.21: 34364023-34364023 on Build GRCh38
rs1025676765 Chr.21: 34364397-34364397 on Build GRCh38
rs41260744 Chr.21: 34364085-34364085 on Build GRCh38
rs72550226 Chr.21: 34364134-34364134 on Build GRCh38
rs929892781 Chr.21: 34364058-34364058 on Build GRCh38
rs940534178 Chr.21: 34364193-34364193 on Build GRCh38
rs1053715725 Chr.21: 34364111-34364111 on Build GRCh38
rs41314699 Chr.21: 34364112-34364112 on Build GRCh38
rs188625398 Chr.21: 34364043-34364043 on Build GRCh38
rs978187738 Chr.21: 34364078-34364078 on Build GRCh38
rs553822163 Chr.21: 34364415-34364415 on Build GRCh38
rs577517827 Chr.21: 34364380-34364380 on Build GRCh38
rs41315511 Chr.21: 34364079-34364079 on Build GRCh38
rs552549792 Chr.21: 34364271-34364272 on Build GRCh38

View Full Product Details