Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020232.4 | NP_064617.2 | 6 - 7 | 827 | 93 |
NM_147163.1 | NP_671692.1 | 6 - 7 | 685 | 93 | |
GenBank mRNA | AF068295.1 | - | 6 - 7 | 685 | 93 |
AF168712.1 | - | 6 - 7 | 759 | 93 | |
AF276707.1 | - | 6 - 7 | 1387 | 93 | |
AK025410.1 | - | 2 - 3 | 173 | 93 | |
AK057005.1 | - | 6 - 7 | 759 | 93 | |
BC013356.2 | - | 6 - 7 | 750 | 93 | |
CR457181.1 | - | 5 - 6 | 587 | 93 |