Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021090.3 | NP_066576.1 | 7 - 8 | 787 | 97 |
NM_153050.2 | NP_694690.1 | 7 - 8 | 787 | 97 | |
NM_153051.2 | NP_694691.1 | 7 - 8 | 787 | 97 | |
XM_005261803.2 | XP_005261860.1 | 8 - 9 | 905 | 97 | |
XM_005261804.3 | XP_005261861.1 | 7 - 8 | 822 | 97 | |
XM_005261805.2 | XP_005261862.1 | 8 - 9 | 905 | 97 | |
XM_005261806.2 | XP_005261863.1 | 7 - 8 | 787 | 97 | |
XM_005261808.2 | XP_005261865.1 | 8 - 9 | 905 | 97 | |
XM_017029028.1 | XP_016884517.1 | 6 - 7 | 679 | 97 | |
XM_017029029.1 | XP_016884518.1 | 6 - 7 | 669 | 97 | |
XM_017029030.1 | XP_016884519.1 | 6 - 7 | 685 | 97 | |
XM_017029031.1 | XP_016884520.1 | 6 - 7 | 673 | 97 | |
XM_017029032.1 | XP_016884521.1 | 7 - 8 | 670 | 97 | |
XM_017029034.1 | XP_016884523.1 | 7 - 8 | 760 | 97 | |
GenBank mRNA | AB002369.1 | - | 7 - 8 | 706 | 97 |
AF233436.2 | - | 4 - 5 | 468 | 97 | |
AF233437.1 | - | 4 - 5 | 468 | 97 | |
AF233438.1 | - | 4 - 5 | 468 | 97 | |
AK289581.1 | - | 7 - 8 | 787 | 97 | |
BC142713.1 | - | 7 - 8 | 743 | 97 | |
BC148216.1 | - | 6 - 7 | 606 | 97 | |
BC151217.1 | - | 7 - 8 | 801 | 97 | |
BC152455.1 | - | 7 - 8 | 706 | 97 | |
CR456525.1 | - | 5 - 6 | 483 | 97 |