Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021167.4 | NP_066990.3 | 4 - 5 | 903 | 85 |
NR_052016.1 | 4 - 5 | 903 | 85 | ||
GenBank mRNA | AB073650.1 | - | 2 - 3 | 997 | 85 |
AK026142.1 | - | 4 - 5 | 872 | 85 | |
AK098014.1 | - | 2 - 3 | 1229 | 85 | |
AK315788.1 | - | 4 - 5 | 869 | 85 | |
BC019350.1 | - | 4 - 5 | 874 | 85 | |
BC031091.1 | - | 4 - 5 | 777 | 85 |