Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021245.3 | NP_067068.1 | 2 - 3 | 440 | 86 |
GenBank mRNA | AF240633.1 | - | 2 - 3 | 142 | 86 |
AJ278124.1 | - | 2 - 3 | 433 | 86 | |
AK300683.1 | - | 2 - 3 | 197 | 86 | |
AK308200.1 | - | 2 - 3 | 197 | 86 | |
AL359210.1 | - | 2 - 3 | 129 | 86 | |
AY013297.1 | - | 2 - 3 | 162 | 86 | |
BC025753.1 | - | 2 - 3 | 440 | 86 |