Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316887.1 | NP_001303816.1 | 3 - 4 | 245 | 81 |
NM_001316888.1 | NP_001303817.1 | 2 - 3 | 273 | 81 | |
NM_021796.3 | NP_068568.1 | 2 - 3 | 227 | 81 | |
XM_011531257.2 | XP_011529559.1 | 2 - 3 | 153 | 81 | |
XM_011531260.2 | XP_011529562.1 | 2 - 3 | 267 | 81 | |
GenBank mRNA | AF234654.1 | - | 2 - 3 | 232 | 81 |
AK075086.1 | - | 2 - 3 | 227 | 81 | |
BC022335.1 | - | 2 - 3 | 212 | 81 |