Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021930.4 | NP_068749.3 | 10 - 11 | 1696 | 76 |
XM_005250524.3 | XP_005250581.1 | 6 - 7 | 878 | 76 | |
XM_011516458.2 | XP_011514760.1 | 6 - 7 | 851 | 76 | |
XM_017012496.1 | XP_016867985.1 | 10 - 11 | 1475 | 76 | |
XM_017012497.1 | XP_016867986.1 | 10 - 11 | 1567 | 76 | |
XM_017012498.1 | XP_016867987.1 | 7 - 8 | 1090 | 76 | |
GenBank mRNA | AF317622.1 | - | 10 - 11 | 1575 | 76 |
AK021847.1 | - | 2 - 3 | 368 | 76 | |
AK055508.1 | - | 10 - 11 | 1611 | 76 | |
AK057757.1 | - | 7 - 8 | 1097 | 76 | |
AK308126.1 | - | 5 - 6 | 489 | 76 | |
AL832825.1 | - | 4 - 5 | 524 | 76 | |
BC068483.1 | - | 10 - 11 | 1697 | 76 |