Assay Design



Gene Transcripts

Gene Symbol
PBLD
Entrez Gene ID
64081
Gene Name
phenazine biosynthesis like protein domain containing
Gene Aliases
HEL-S-306, MAWBP, MAWDBP
Chromosome Location
Chr.10: 68282660 - 68332958 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033083.1 NP_001028255.1 8 - 9 896 65
NM_022129.3 NP_071412.2 8 - 9 896 65
XM_005270028.3 XP_005270085.1 8 - 9 930 65
GenBank mRNA AB049758.1 - 6 - 7 758 65
AI114748.1 - 5 - 6 431 65
AI439144.1 - 1 - 2 65
AK312275.1 - 7 - 8 687 65
BC009738.1 - 8 - 9 878 65
GQ472233.1 - 8 - 9 878 65

View Full Product Details