Assay Design



Gene Transcripts

Gene Symbol
C11orf24
Entrez Gene ID
53838
Gene Name
chromosome 11 open reading frame 24
Gene Aliases
DM4E3
Chromosome Location
Chr.11: 68261335 - 68272001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300913.1 NP_001287842.1 1 - 2 145 108
NM_022338.3 NP_071733.1 1 - 2 145 108
GenBank mRNA AF264781.1 - 1 - 2 105 108
BG701401.1 - 1 - 2 150 108
BI862360.1 - 1 - 2 119 108
DB471655.1 - 1 - 2 145 108

View Full Product Details