Assay Design



Gene Transcripts

Gene Symbol
DGCR14
Entrez Gene ID
8220
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el
Chromosome Location
Chr.22: 19130279 - 19144726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022719.2 NP_073210.1 8 - 9 1073 109
NR_134304.1 8 - 9 1150 109
XM_005261282.4 XP_005261339.1 8 - 9 1081 109
XM_006724329.3 XP_006724392.1 8 - 9 1440 109
XM_006724330.3 XP_006724393.1 8 - 9 1481 109
XM_006724331.3 XP_006724394.1 8 - 9 1478 109
XM_017028958.1 XP_016884447.1 8 - 9 1437 109
XM_017028959.1 XP_016884448.1 2 - 3 541 109
GenBank mRNA BC003015.1 - 8 - 9 1035 109
BC006542.2 - 8 - 9 1037 109
BX428458.2 - 3 - 4 424 109
CR456344.1 - 8 - 9 1045 109
L77566.1 - 8 - 9 1046 109
L78010.1 - 8 - 9 1042 109

View Full Product Details