Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022819.3 | NP_073730.3 | 2 - 3 | 271 | 70 |
XM_011541955.1 | XP_011540257.1 | 2 - 3 | 264 | 70 | |
XM_011541956.1 | XP_011540258.1 | 2 - 3 | 275 | 70 | |
GenBank mRNA | AF306566.1 | - | 1 - 2 | 44 | 70 |
AK093645.1 | - | 2 - 3 | 275 | 70 |