Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38112113-38112603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G6
Entrez Gene ID
8398
Gene Name
phospholipase A2 group VI
Gene Aliases
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
UniGene
Hs.170479
Gene Chromosome Location
Chr.22: 38111495-38192099 on Build GRCh38
Transcripts
NM_001004426.1 NM_001199562.1 NM_003560.2 XM_005261764.2 XM_005261765.1 XM_005261766.1 XM_006724332.3 XM_011530422.1 XM_011530423.1 XM_011530424.1 XM_011530425.1 XM_017028981.1 XM_017028982.1 XM_017028983.1 XM_017028984.1 XM_017028985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5523497 c.2367C>T p.I789I Chr.22: 38112215-38112215 on Build GRCh38
4388868 c.2212C>A p.P738T Chr.22: 38112568-38112568 on Build GRCh38
6095375 c.2412C>T p.L804L Chr.22: 38112170-38112170 on Build GRCh38
3713734 c.2299G>C p.D767H Chr.22: 38112283-38112283 on Build GRCh38
6499871 c.2265C>G p.I755M Chr.22: 38112515-38112515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963927447 Chr.22: 38112520-38112520 on Build GRCh38
rs530351387 Chr.22: 38112204-38112204 on Build GRCh38
rs373493102 Chr.22: 38112295-38112295 on Build GRCh38
rs766616183 Chr.22: 38112284-38112284 on Build GRCh38
rs370528124 Chr.22: 38112311-38112311 on Build GRCh38
rs548439730 Chr.22: 38112563-38112563 on Build GRCh38
rs761294759 Chr.22: 38112491-38112491 on Build GRCh38
rs121908687 Chr.22: 38112541-38112541 on Build GRCh38
rs756904560 Chr.22: 38112223-38112223 on Build GRCh38
rs201208246 Chr.22: 38112285-38112285 on Build GRCh38
rs530348521 Chr.22: 38112559-38112559 on Build GRCh38
rs11570766 Chr.22: 38112372-38112372 on Build GRCh38
rs1043504064 Chr.22: 38112399-38112399 on Build GRCh38
rs909426902 Chr.22: 38112145-38112146 on Build GRCh38
rs587784351 Chr.22: 38112534-38112534 on Build GRCh38
rs748913525 Chr.22: 38112143-38112143 on Build GRCh38
rs776793367 Chr.22: 38112198-38112198 on Build GRCh38
rs796331166 Chr.22: 38112405-38112405 on Build GRCh38
rs139093920 Chr.22: 38112241-38112241 on Build GRCh38
rs749437245 Chr.22: 38112462-38112462 on Build GRCh38
rs924915026 Chr.22: 38112404-38112404 on Build GRCh38
rs544263173 Chr.22: 38112527-38112527 on Build GRCh38
rs551260190 Chr.22: 38112141-38112141 on Build GRCh38
rs955215757 Chr.22: 38112351-38112351 on Build GRCh38
rs554827625 Chr.22: 38112455-38112455 on Build GRCh38
rs910985603 Chr.22: 38112147-38112147 on Build GRCh38
rs377191625 Chr.22: 38112142-38112142 on Build GRCh38
rs1030901001 Chr.22: 38112320-38112320 on Build GRCh38
rs571223704 Chr.22: 38112312-38112312 on Build GRCh38
rs34184838 Chr.22: 38112261-38112261 on Build GRCh38
rs267606245 Chr.22: 38112289-38112289 on Build GRCh38
rs11570764 Chr.22: 38112461-38112461 on Build GRCh38
rs587784354 Chr.22: 38112186-38112186 on Build GRCh38
rs770983765 Chr.22: 38112572-38112572 on Build GRCh38
rs773403661 Chr.22: 38112276-38112276 on Build GRCh38
rs182038896 Chr.22: 38112415-38112415 on Build GRCh38
rs775266092 Chr.22: 38112176-38112176 on Build GRCh38
rs138683183 Chr.22: 38112242-38112242 on Build GRCh38
rs41274545 Chr.22: 38112361-38112361 on Build GRCh38
rs756816515 Chr.22: 38112308-38112308 on Build GRCh38
rs770730885 Chr.22: 38112162-38112162 on Build GRCh38
rs778705470 Chr.22: 38112226-38112226 on Build GRCh38
rs149605475 Chr.22: 38112213-38112213 on Build GRCh38
rs933380615 Chr.22: 38112201-38112201 on Build GRCh38
rs772143897 Chr.22: 38112571-38112571 on Build GRCh38
rs771731816 Chr.22: 38112228-38112228 on Build GRCh38
rs764789953 Chr.22: 38112519-38112519 on Build GRCh38
rs774621327 Chr.22: 38112580-38112580 on Build GRCh38
rs949569745 Chr.22: 38112447-38112447 on Build GRCh38
rs751982449 Chr.22: 38112206-38112206 on Build GRCh38
rs779749576 Chr.22: 38112530-38112530 on Build GRCh38
rs765446322 Chr.22: 38112203-38112203 on Build GRCh38
rs565332748 Chr.22: 38112467-38112467 on Build GRCh38
rs150093048 Chr.22: 38112215-38112215 on Build GRCh38
rs759942729 Chr.22: 38112485-38112485 on Build GRCh38
rs749966284 Chr.22: 38112521-38112521 on Build GRCh38
rs376144077 Chr.22: 38112540-38112540 on Build GRCh38
rs764039953 Chr.22: 38112188-38112188 on Build GRCh38
rs121908680 Chr.22: 38112212-38112212 on Build GRCh38
rs868205860 Chr.22: 38112172-38112172 on Build GRCh38
rs374303522 Chr.22: 38112474-38112474 on Build GRCh38
rs760705741 Chr.22: 38112181-38112181 on Build GRCh38
rs761874020 Chr.22: 38112266-38112266 on Build GRCh38
rs746013568 Chr.22: 38112562-38112562 on Build GRCh38
rs35537874 Chr.22: 38112164-38112164 on Build GRCh38
rs765356472 Chr.22: 38112274-38112274 on Build GRCh38
rs375779632 Chr.22: 38112472-38112472 on Build GRCh38
rs986738833 Chr.22: 38112481-38112481 on Build GRCh38
rs587784352 Chr.22: 38112254-38112255 on Build GRCh38
rs761961717 Chr.22: 38112202-38112202 on Build GRCh38
rs563222818 Chr.22: 38112544-38112544 on Build GRCh38
rs993482020 Chr.22: 38112468-38112468 on Build GRCh38
rs775386225 Chr.22: 38112233-38112233 on Build GRCh38
rs745664459 Chr.22: 38112166-38112166 on Build GRCh38
rs778604977 Chr.22: 38112309-38112309 on Build GRCh38
rs372267803 Chr.22: 38112490-38112490 on Build GRCh38
rs914530056 Chr.22: 38112402-38112402 on Build GRCh38
rs11570765 Chr.22: 38112434-38112434 on Build GRCh38
rs1045677686 Chr.22: 38112452-38112452 on Build GRCh38
rs34910119 Chr.22: 38112371-38112372 on Build GRCh38
rs587784349 Chr.22: 38112565-38112565 on Build GRCh38
rs766818779 Chr.22: 38112205-38112205 on Build GRCh38
rs368514303 Chr.22: 38112546-38112546 on Build GRCh38
rs571458275 Chr.22: 38112457-38112457 on Build GRCh38
rs980430577 Chr.22: 38112582-38112582 on Build GRCh38
rs140758033 Chr.22: 38112165-38112165 on Build GRCh38
rs747612137 Chr.22: 38112138-38112138 on Build GRCh38
rs587784353 Chr.22: 38112211-38112212 on Build GRCh38
rs376089103 Chr.22: 38112240-38112240 on Build GRCh38
rs577215031 Chr.22: 38112487-38112487 on Build GRCh38
rs368835758 Chr.22: 38112489-38112489 on Build GRCh38
rs886184330 Chr.22: 38112449-38112449 on Build GRCh38
rs121908686 Chr.22: 38112558-38112558 on Build GRCh38
rs893021938 Chr.22: 38112207-38112207 on Build GRCh38
rs938490717 Chr.22: 38112443-38112443 on Build GRCh38
rs753256678 Chr.22: 38112375-38112375 on Build GRCh38
rs751225193 Chr.22: 38112531-38112531 on Build GRCh38
rs754070657 Chr.22: 38112358-38112358 on Build GRCh38
rs768074348 Chr.22: 38112297-38112297 on Build GRCh38
rs373689745 Chr.22: 38112458-38112458 on Build GRCh38
rs374816889 Chr.22: 38112314-38112314 on Build GRCh38
rs763205259 Chr.22: 38112277-38112277 on Build GRCh38
rs370465424 Chr.22: 38112471-38112471 on Build GRCh38
rs772061693 Chr.22: 38112175-38112175 on Build GRCh38
rs587784350 Chr.22: 38112547-38112547 on Build GRCh38
rs748821599 Chr.22: 38112218-38112218 on Build GRCh38
rs936682716 Chr.22: 38112388-38112388 on Build GRCh38
rs552606315 Chr.22: 38112307-38112307 on Build GRCh38
rs146591896 Chr.22: 38112227-38112227 on Build GRCh38

View Full Product Details