Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38129365-38129865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G6
Entrez Gene ID
8398
Gene Name
phospholipase A2 group VI
Gene Aliases
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
UniGene
Hs.170479
Gene Chromosome Location
Chr.22: 38111495-38192099 on Build GRCh38
Transcripts
NM_001004426.1 NM_001199562.1 NM_003560.2 XM_005261764.2 XM_005261765.1 XM_005261766.1 XM_006724332.3 XM_011530422.1 XM_011530423.1 XM_011530424.1 XM_011530425.1 XM_011530426.2 XM_017028981.1 XM_017028982.1 XM_017028983.1 XM_017028984.1 XM_017028985.1 XM_017028986.1 XM_017028987.1 XM_017028988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3151148 c.1185A>G p.R395R Chr.22: 38129455-38129455 on Build GRCh38
1034106 c.1179C>T p.I393I Chr.22: 38129461-38129461 on Build GRCh38
5592556 c.973-1G>A p.? Chr.22: 38129563-38129563 on Build GRCh38
6506270 c.1037A>G p.D346G Chr.22: 38129498-38129498 on Build GRCh38
1714385 c.1136C>G p.P379R Chr.22: 38129504-38129504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756375070 Chr.22: 38129575-38129576 on Build GRCh38
rs761082713 Chr.22: 38129601-38129601 on Build GRCh38
rs756615798 Chr.22: 38129541-38129541 on Build GRCh38
rs766618805 Chr.22: 38129520-38129520 on Build GRCh38
rs748042748 Chr.22: 38129582-38129582 on Build GRCh38
rs190185377 Chr.22: 38129589-38129589 on Build GRCh38
rs769930709 Chr.22: 38129507-38129507 on Build GRCh38
rs921559500 Chr.22: 38129444-38129444 on Build GRCh38
rs772304837 Chr.22: 38129395-38129395 on Build GRCh38
rs781641825 Chr.22: 38129440-38129440 on Build GRCh38
rs774582699 Chr.22: 38129594-38129594 on Build GRCh38
rs376497527 Chr.22: 38129537-38129537 on Build GRCh38
rs562743092 Chr.22: 38129807-38129807 on Build GRCh38
rs982579080 Chr.22: 38129678-38129678 on Build GRCh38
rs982464698 Chr.22: 38129396-38129396 on Build GRCh38
rs773091386 Chr.22: 38129583-38129583 on Build GRCh38
rs965086820 Chr.22: 38129689-38129689 on Build GRCh38
rs371593013 Chr.22: 38129490-38129490 on Build GRCh38
rs1002237537 Chr.22: 38129584-38129584 on Build GRCh38
rs538112875 Chr.22: 38129524-38129524 on Build GRCh38
rs750742993 Chr.22: 38129435-38129436 on Build GRCh38
rs144041114 Chr.22: 38129695-38129695 on Build GRCh38
rs1045647122 Chr.22: 38129779-38129779 on Build GRCh38
rs769061741 Chr.22: 38129585-38129585 on Build GRCh38
rs906644051 Chr.22: 38129825-38129825 on Build GRCh38
rs775654871 Chr.22: 38129437-38129437 on Build GRCh38
rs759783011 Chr.22: 38129595-38129595 on Build GRCh38
rs943747232 Chr.22: 38129432-38129432 on Build GRCh38
rs775059242 Chr.22: 38129472-38129472 on Build GRCh38
rs972262861 Chr.22: 38129482-38129482 on Build GRCh38
rs746612515 Chr.22: 38129730-38129730 on Build GRCh38
rs768535131 Chr.22: 38129577-38129577 on Build GRCh38
rs149554238 Chr.22: 38129545-38129545 on Build GRCh38
rs776664421 Chr.22: 38129503-38129503 on Build GRCh38
rs765156550 Chr.22: 38129529-38129529 on Build GRCh38
rs181622396 Chr.22: 38129699-38129699 on Build GRCh38
rs554347201 Chr.22: 38129600-38129600 on Build GRCh38
rs564845293 Chr.22: 38129661-38129661 on Build GRCh38
rs751940622 Chr.22: 38129429-38129429 on Build GRCh38
rs575898460 Chr.22: 38129461-38129461 on Build GRCh38
rs936951867 Chr.22: 38129724-38129724 on Build GRCh38
rs756705763 Chr.22: 38129447-38129447 on Build GRCh38
rs540249986 Chr.22: 38129646-38129646 on Build GRCh38
rs778225931 Chr.22: 38129543-38129543 on Build GRCh38
rs770965124 Chr.22: 38129590-38129590 on Build GRCh38
rs746792356 Chr.22: 38129473-38129473 on Build GRCh38
rs767889564 Chr.22: 38129530-38129530 on Build GRCh38
rs555096269 Chr.22: 38129800-38129800 on Build GRCh38
rs914075240 Chr.22: 38129753-38129753 on Build GRCh38
rs897702872 Chr.22: 38129784-38129784 on Build GRCh38
rs587784327 Chr.22: 38129523-38129523 on Build GRCh38
rs758653527 Chr.22: 38129424-38129424 on Build GRCh38
rs767024328 Chr.22: 38129781-38129781 on Build GRCh38
rs761815070 Chr.22: 38129453-38129453 on Build GRCh38
rs778423680 Chr.22: 38129454-38129454 on Build GRCh38
rs2413502 Chr.22: 38129554-38129554 on Build GRCh38
rs1019358390 Chr.22: 38129687-38129687 on Build GRCh38
rs535278511 Chr.22: 38129602-38129602 on Build GRCh38
rs113789271 Chr.22: 38129709-38129709 on Build GRCh38
rs746666741 Chr.22: 38129565-38129565 on Build GRCh38
rs11570693 Chr.22: 38129623-38129623 on Build GRCh38
rs754574930 Chr.22: 38129732-38129732 on Build GRCh38
rs773356186 Chr.22: 38129510-38129510 on Build GRCh38
rs745443102 Chr.22: 38129460-38129460 on Build GRCh38
rs1033599478 Chr.22: 38129387-38129387 on Build GRCh38
rs371925461 Chr.22: 38129823-38129823 on Build GRCh38
rs556727891 Chr.22: 38129532-38129532 on Build GRCh38
rs763001695 Chr.22: 38129514-38129514 on Build GRCh38
rs570834054 Chr.22: 38129504-38129504 on Build GRCh38
rs1014609659 Chr.22: 38129710-38129710 on Build GRCh38
rs765433063 Chr.22: 38129415-38129415 on Build GRCh38
rs752076777 Chr.22: 38129844-38129844 on Build GRCh38
rs760467408 Chr.22: 38129406-38129406 on Build GRCh38
rs776753734 Chr.22: 38129409-38129409 on Build GRCh38
rs951179982 Chr.22: 38129675-38129675 on Build GRCh38
rs761965546 Chr.22: 38129411-38129411 on Build GRCh38
rs748701861 Chr.22: 38129446-38129446 on Build GRCh38
rs764653387 Chr.22: 38129604-38129604 on Build GRCh38
rs781141708 Chr.22: 38129578-38129578 on Build GRCh38
rs750573751 Chr.22: 38129422-38129422 on Build GRCh38
rs764105141 Chr.22: 38129407-38129407 on Build GRCh38
rs144168978 Chr.22: 38129553-38129553 on Build GRCh38

View Full Product Details