Product Details

Assay Reference Genome
Location

Chr.11:72232709 on build GRCh38
Cytoband
11q13.4
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
3636
Gene Symbol
INPPL1
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
Location
Chr.11:72223861-72239147 on build GRCh38
Assay Gene Location
Within Exon 17
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
INPPL1 NM_001567.3 15 1943 NP_001558.3
XM_005273978.3 16 2320 XP_005274035.1
XM_005273979.4 16 2167 XP_005274036.1
XM_011544999.2 16 2101 XP_011543301.1
XM_011545000.2 16 2167 XP_011543302.1
XM_017017673.1 17 2258 XP_016873162.1
BC140853.1 15 1943
L36818.1 14 1920 AAA96658.1
Y14385.1 15 2007 CAA74743.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv951038 Chr11:72226157 - 72253456 on Build GRCh38 Deletion LOC220077 PHOX2A INPPL1

View Full Product Details