Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319952.1 | NP_001306881.1 | 11 - 12 | 1293 | 132 |
NM_024526.3 | NP_078802.2 | 12 - 13 | 1345 | 132 | |
NM_133181.3 | NP_573444.2 | 12 - 13 | 1345 | 132 | |
NM_139053.2 | NP_620641.1 | 12 - 13 | 1348 | 132 | |
XM_011542132.1 | XP_011540434.1 | 12 - 13 | 1270 | 132 | |
XM_011542133.1 | XP_011540435.1 | 12 - 13 | 1267 | 132 | |
XM_011542134.2 | XP_011540436.1 | 11 - 12 | 1269 | 132 | |
XM_011542135.2 | XP_011540437.1 | 12 - 13 | 1387 | 132 | |
XM_017002327.1 | XP_016857816.1 | 12 - 13 | 1270 | 132 | |
XM_017002328.1 | XP_016857817.1 | 12 - 13 | 1270 | 132 | |
XM_017002329.1 | XP_016857818.1 | 12 - 13 | 1246 | 132 | |
GenBank mRNA | AK025175.1 | - | 12 - 13 | 1247 | 132 |
AK225235.1 | - | 12 - 13 | 1247 | 132 | |
AK290261.1 | - | 12 - 13 | 1272 | 132 | |
AK292198.1 | - | 12 - 13 | 1269 | 132 | |
AK307932.1 | - | 11 - 12 | 1191 | 132 | |
AK308196.1 | - | 11 - 12 | 1087 | 132 | |
AK310979.1 | - | 12 - 13 | 1332 | 132 | |
AY074930.1 | - | 12 - 13 | 1231 | 132 | |
BC012926.1 | - | 12 - 13 | 1268 | 132 | |
BP419077.1 | - | 4 - 5 | 341 | 132 |