Assay Design



Gene Transcripts

Gene Symbol
TCTN1
Entrez Gene ID
79600
Gene Name
tectonic family member 1
Gene Aliases
JBTS13, TECT1
Chromosome Location
Chr.12: 110614027 - 110649130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082537.2 NP_001076006.1 2 - 3 496 92
NM_001082538.2 NP_001076007.1 2 - 3 496 92
NM_001173975.2 NP_001167446.1 2 - 3 557 92
NM_001173976.1 NP_001167447.1 3 - 4 572 92
NM_001319680.1 NP_001306609.1 2 - 3 496 92
NM_001319681.1 NP_001306610.1 2 - 3 496 92
NM_001319682.1 NP_001306611.1 2 - 3 557 92
NM_024549.5 NP_078825.2 2 - 3 496 92
NR_135088.1 2 - 3 557 92
XM_005253934.3 XP_005253991.1 2 - 3 400 92
XM_005253935.3 XP_005253992.1 2 - 3 400 92
XM_006719594.2 XP_006719657.1 2 - 3 461 92
XM_006719595.2 XP_006719658.1 3 - 4 567 92
XM_006719596.2 XP_006719659.1 2 - 3 417 92
XM_006719598.2 XP_006719661.1 2 - 3 417 92
XM_006719599.2 XP_006719662.1 2 - 3 395 92
XM_011538733.2 XP_011537035.1 2 - 3 400 92
XM_011538735.1 XP_011537037.1 2 - 3 400 92
XM_011538737.2 XP_011537039.1 2 - 3 400 92
XM_011538738.2 XP_011537040.1 2 - 3 400 92
XM_017019964.1 XP_016875453.1 2 - 3 461 92
XM_017019965.1 XP_016875454.1 2 - 3 384 92
XM_017019966.1 XP_016875455.1 2 - 3 431 92
XM_017019967.1 XP_016875456.1 2 - 3 261 92
XM_017019969.1 XP_016875458.1 2 - 3 395 92
GenBank mRNA AK024780.1 - 3 - 4 470 92
AK055891.1 - 2 - 3 410 92
AK092775.1 - 2 - 3 458 92
AK295514.1 - 2 - 3 496 92
AK301732.1 - 2 - 3 361 92
AY358184.1 - 2 - 3 359 92
BC033811.1 - 2 - 3 443 92
BC040113.1 - 2 - 3 386 92
BC044885.1 - 2 - 3 419 92
BC062611.1 - 2 - 3 347 92
DN997186.1 - 2 - 3 456 92
DQ278868.1 - 2 - 3 421 92

View Full Product Details