Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001302084.1 | NP_001289013.1 | 4 - 5 | 434 | 136 |
NM_024650.3 | NP_078926.3 | 6 - 7 | 726 | 136 | |
GenBank mRNA | AK026184.1 | - | 5 - 6 | 527 | 136 |
AK225435.1 | - | 5 - 6 | 527 | 136 | |
AK302025.1 | - | 4 - 5 | 386 | 136 | |
BC028240.1 | - | 6 - 7 | 729 | 136 |