Assay Design



Gene Transcripts

Gene Symbol
THAP9
Entrez Gene ID
79725
Gene Name
THAP domain containing 9
Gene Aliases
hTh9
Chromosome Location
Chr.4: 82900650 - 82920283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317776.1 NP_001304705.1 5 - 6 951 57
NM_024672.5 NP_078948.3 4 - 5 885 57
XM_005263220.2 XP_005263277.1 4 - 5 842 57
XM_017008603.1 XP_016864092.1 4 - 5 1725 57
XM_017008604.1 XP_016864093.1 4 - 5 1778 57
XM_017008605.1 XP_016864094.1 4 - 5 819 57
XM_017008606.1 XP_016864095.1 5 - 6 943 57
XM_017008607.1 XP_016864096.1 4 - 5 1158 57
XM_017008608.1 XP_016864097.1 5 - 6 1797 57
GenBank mRNA AK026973.1 - 2 - 3 215 57
AK091412.1 - 4 - 5 782 57
AK225333.1 - 2 - 3 222 57
BC131722.1 - 5 - 6 832 57
DB154392.1 - 2 - 3 360 57

View Full Product Details