Assay Design



Gene Transcripts

Gene Symbol
C16orf95
Entrez Gene ID
100506581
Gene Name
chromosome 16 open reading frame 95
Gene Aliases
-
Chromosome Location
Chr.16: 87302798 - 87317420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195125.1 NP_001182054.1 3 - 4 508 71
NM_001256917.1 NP_001243846.1 3 - 4 512 71
GenBank mRNA AB371437.1 - 3 - 4 480 71
AK026130.1 - 3 - 4 504 71
BC015536.2 - 3 - 4 448 71
BI460222.1 - 3 - 4 511 71
BU839368.1 - 2 - 3 71
DA871621.1 - 3 - 4 472 71

View Full Product Details