Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024764.3 | NP_079040.2 | 17 - 18 | 1902 | 102 |
GenBank mRNA | AK024360.1 | - | 3 - 4 | 430 | 102 |
AK126034.1 | - | 17 - 18 | 1886 | 102 | |
BC126214.1 | - | 17 - 18 | 1856 | 102 | |
BC126216.1 | - | 17 - 18 | 1856 | 102 |