Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14132775-14133272 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
UniGene
Hs.517817
Gene Chromosome Location
Chr.3: 14124940-14143680 on Build GRCh38
Transcripts
NM_024334.2 XM_017007176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1419494 c.483C>T p.F161F Chr.3: 14132906-14132906 on Build GRCh38
728996 c.497G>T p.G166V Chr.3: 14132920-14132920 on Build GRCh38
5982868 c.488G>A p.R163Q Chr.3: 14132911-14132911 on Build GRCh38
326696 c.499C>T p.H167Y Chr.3: 14132922-14132922 on Build GRCh38
3759801 c.504A>T p.K168N Chr.3: 14132927-14132927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961847604 Chr.3: 14132848-14132848 on Build GRCh38
rs74553717 Chr.3: 14133061-14133061 on Build GRCh38
rs772312969 Chr.3: 14132980-14132980 on Build GRCh38
rs892672013 Chr.3: 14133030-14133030 on Build GRCh38
rs374494474 Chr.3: 14133054-14133054 on Build GRCh38
rs766205020 Chr.3: 14132947-14132947 on Build GRCh38
rs759866439 Chr.3: 14133211-14133211 on Build GRCh38
rs370539390 Chr.3: 14132820-14132820 on Build GRCh38
rs777743046 Chr.3: 14132961-14132961 on Build GRCh38
rs142500068 Chr.3: 14132819-14132819 on Build GRCh38
rs114026215 Chr.3: 14132954-14132954 on Build GRCh38
rs941251363 Chr.3: 14133187-14133187 on Build GRCh38
rs747270926 Chr.3: 14133159-14133159 on Build GRCh38
rs794729181 Chr.3: 14132932-14132932 on Build GRCh38
rs368256924 Chr.3: 14132817-14132817 on Build GRCh38
rs914115298 Chr.3: 14133226-14133227 on Build GRCh38
rs1051083755 Chr.3: 14133228-14133228 on Build GRCh38
rs539231825 Chr.3: 14133098-14133098 on Build GRCh38
rs575962784 Chr.3: 14133198-14133198 on Build GRCh38
rs981819676 Chr.3: 14133082-14133082 on Build GRCh38
rs769654404 Chr.3: 14132891-14132891 on Build GRCh38
rs1019702462 Chr.3: 14132800-14132800 on Build GRCh38
rs767516734 Chr.3: 14132920-14132920 on Build GRCh38
rs757632483 Chr.3: 14132943-14132943 on Build GRCh38
rs776246023 Chr.3: 14133207-14133207 on Build GRCh38
rs916845585 Chr.3: 14132985-14132985 on Build GRCh38
rs758030788 Chr.3: 14132834-14132834 on Build GRCh38
rs772842801 Chr.3: 14132906-14132906 on Build GRCh38
rs769722980 Chr.3: 14132904-14132904 on Build GRCh38
rs780800552 Chr.3: 14132955-14132955 on Build GRCh38
rs750107818 Chr.3: 14132832-14132832 on Build GRCh38
rs531528770 Chr.3: 14133077-14133077 on Build GRCh38
rs770753162 Chr.3: 14132977-14132977 on Build GRCh38
rs373229565 Chr.3: 14132837-14132837 on Build GRCh38
rs775986881 Chr.3: 14132923-14132923 on Build GRCh38
rs969051076 Chr.3: 14133074-14133074 on Build GRCh38
rs4685076 Chr.3: 14132927-14132927 on Build GRCh38
rs865789335 Chr.3: 14133068-14133068 on Build GRCh38
rs746579064 Chr.3: 14132841-14132841 on Build GRCh38
rs989785149 Chr.3: 14133215-14133215 on Build GRCh38
rs1039635203 Chr.3: 14133192-14133192 on Build GRCh38
rs965409708 Chr.3: 14132809-14132809 on Build GRCh38
rs1024692757 Chr.3: 14133191-14133191 on Build GRCh38
rs756046294 Chr.3: 14132960-14132960 on Build GRCh38
rs929744688 Chr.3: 14133138-14133138 on Build GRCh38
rs759676847 Chr.3: 14132911-14132911 on Build GRCh38
rs868824693 Chr.3: 14133033-14133033 on Build GRCh38
rs761672806 Chr.3: 14132832-14132832 on Build GRCh38
rs368761778 Chr.3: 14133225-14133225 on Build GRCh38
rs768069110 Chr.3: 14132823-14132823 on Build GRCh38
rs1028299336 Chr.3: 14133075-14133075 on Build GRCh38
rs756093189 Chr.3: 14132829-14132829 on Build GRCh38
rs949064136 Chr.3: 14132915-14132915 on Build GRCh38
rs1006172546 Chr.3: 14133111-14133111 on Build GRCh38
rs748042640 Chr.3: 14132890-14132890 on Build GRCh38
rs901222045 Chr.3: 14133064-14133064 on Build GRCh38
rs957839594 Chr.3: 14133042-14133042 on Build GRCh38
rs1046932119 Chr.3: 14132866-14132866 on Build GRCh38
rs888352687 Chr.3: 14132895-14132895 on Build GRCh38
rs756472571 Chr.3: 14132826-14132826 on Build GRCh38
rs980913166 Chr.3: 14133195-14133195 on Build GRCh38
rs888426020 Chr.3: 14133084-14133084 on Build GRCh38
rs111341618 Chr.3: 14132824-14132824 on Build GRCh38
rs764546967 Chr.3: 14132936-14132936 on Build GRCh38
rs771118236 Chr.3: 14133164-14133164 on Build GRCh38
rs559204680 Chr.3: 14133169-14133169 on Build GRCh38
rs773242841 Chr.3: 14133037-14133037 on Build GRCh38
rs777049359 Chr.3: 14133170-14133170 on Build GRCh38
rs147336367 Chr.3: 14132910-14132910 on Build GRCh38
rs546360883 Chr.3: 14133011-14133011 on Build GRCh38
rs754209349 Chr.3: 14132938-14132938 on Build GRCh38
rs368555747 Chr.3: 14132854-14132854 on Build GRCh38
rs1029708362 Chr.3: 14132974-14132974 on Build GRCh38
rs748128413 Chr.3: 14132958-14132958 on Build GRCh38
rs774557631 Chr.3: 14132984-14132984 on Build GRCh38
rs371866082 Chr.3: 14132857-14132857 on Build GRCh38
rs150425166 Chr.3: 14132907-14132907 on Build GRCh38
rs868575020 Chr.3: 14132931-14132931 on Build GRCh38
rs1033211861 Chr.3: 14133038-14133038 on Build GRCh38
rs899807606 Chr.3: 14133197-14133197 on Build GRCh38
rs41284009 Chr.3: 14132818-14132818 on Build GRCh38
rs749093805 Chr.3: 14132963-14132963 on Build GRCh38
rs927697338 Chr.3: 14133116-14133116 on Build GRCh38

View Full Product Details