Assay Design



Gene Transcripts

Gene Symbol
ATP8B4
Entrez Gene ID
79895
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Gene Aliases
ATPIM
Chromosome Location
Chr.15: 49858238 - 50182096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024837.3 NP_079113.2 24 - 25 2925 110
NR_073596.1 24 - 25 2824 110
NR_073597.1 23 - 24 2777 110
NR_073598.1 25 - 26 3032 110
XM_011522046.2 XP_011520348.1 26 - 27 3314 110
XM_011522047.2 XP_011520349.1 25 - 26 3839 110
XM_011522048.1 XP_011520350.1 25 - 26 3112 110
XM_011522049.2 XP_011520351.1 25 - 26 3222 110
XM_011522051.2 XP_011520353.1 26 - 27 3149 110
XM_011522052.2 XP_011520354.1 25 - 26 3207 110
XM_011522053.1 XP_011520355.1 25 - 26 2988 110
XM_011522056.2 XP_011520358.2 25 - 26 3374 110
XM_011522058.2 XP_011520360.1 21 - 22 3583 110
XM_011522059.1 XP_011520361.1 20 - 21 2604 110
XM_011522060.1 XP_011520362.1 23 - 24 2839 110
XM_011522061.1 XP_011520363.1 22 - 23 2754 110
XM_011522062.1 XP_011520364.1 20 - 21 2593 110
XM_011522063.1 XP_011520365.1 20 - 21 2575 110
XM_011522064.1 XP_011520366.1 16 - 17 2276 110
XM_011522069.2 XP_011520371.1 15 - 16 2097 110
XM_011522070.1 XP_011520372.1 11 - 12 1517 110
XM_017022587.1 XP_016878076.1 24 - 25 3290 110
XM_017022588.1 XP_016878077.1 24 - 25 3215 110
XM_017022589.1 XP_016878078.1 24 - 25 3185 110
XM_017022590.1 XP_016878079.1 26 - 27 3126 110
XM_017022591.1 XP_016878080.1 25 - 26 3073 110
XM_017022592.1 XP_016878081.1 24 - 25 3136 110
XM_017022593.1 XP_016878082.1 25 - 26 3374 110
XM_017022594.1 XP_016878083.1 21 - 22 2634 110
XM_017022595.1 XP_016878084.1 21 - 22 4639 110
XM_017022596.1 XP_016878085.1 16 - 17 2267 110
GenBank mRNA AB075819.1 - 25 - 26 3032 110
AK025125.1 - 1 - 2 66 110
AK123747.1 - 5 - 6 691 110
BC144297.1 - 23 - 24 2773 110

View Full Product Details