Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024867.3 | NP_079143.3 | 26 - 27 | 3913 | 70 |
XM_005248376.3 | XP_005248433.1 | 26 - 27 | 4018 | 70 | |
XM_005248377.3 | XP_005248434.1 | 26 - 27 | 4003 | 70 | |
XM_011514135.2 | XP_011512437.1 | 26 - 27 | 4023 | 70 | |
XM_011514136.2 | XP_011512438.1 | 26 - 27 | 4022 | 70 | |
XM_011514137.2 | XP_011512439.1 | 26 - 27 | 4007 | 70 | |
XM_011514138.2 | XP_011512440.1 | 26 - 27 | 3999 | 70 | |
XM_011514139.2 | XP_011512441.1 | 25 - 26 | 3947 | 70 | |
XM_011514140.2 | XP_011512442.1 | 25 - 26 | 3757 | 70 | |
XM_011514141.2 | XP_011512443.1 | 27 - 28 | 4221 | 70 | |
XM_011514142.1 | XP_011512444.1 | 26 - 27 | 3994 | 70 | |
XM_017009880.1 | XP_016865369.1 | 25 - 26 | 3839 | 70 | |
XM_017009881.1 | XP_016865370.1 | 26 - 27 | 4877 | 70 | |
GenBank mRNA | AB051557.1 | - | 16 - 17 | 2254 | 70 |
AK027230.1 | - | 3 - 4 | 359 | 70 | |
AK225586.1 | - | 3 - 4 | 359 | 70 | |
AK225607.1 | - | 3 - 4 | 359 | 70 |