Assay Design



Gene Transcripts

Gene Symbol
DSN1
Entrez Gene ID
79980
Gene Name
DSN1 homolog, MIS12 kinetochore complex component
Gene Aliases
C20orf172, KNL3, MIS13, dJ469A13.2, hKNL-3
Chromosome Location
Chr.20: 36751791 - 36773827 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145315.1 NP_001138787.1 3 - 4 540 74
NM_001145316.1 NP_001138788.1 3 - 4 732 74
NM_001145318.1 NP_001138790.1 2 - 3 491 74
NM_024918.3 NP_079194.3 3 - 4 645 74
XM_017028070.1 XP_016883559.1 3 - 4 528 74
XM_017028071.1 XP_016883560.1 3 - 4 750 74
XM_017028072.1 XP_016883561.1 2 - 3 438 74
GenBank mRNA AK023408.1 - 4 - 5 371 74
AK093031.1 - 3 - 4 640 74
AK290738.1 - 3 - 4 732 74
AK301671.1 - 2 - 3 424 74
BC026011.1 - 2 - 3 745 74
BC058899.1 - 3 - 4 498 74

View Full Product Details